Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or67c
- Entrez Gene ID
-
39189
- Gene Name
- Odorant receptor 67c
- Gene Aliases
-
67D.1, 67c, CG14156, DOR77, DmOR67C, Dmel\CG14156, Dmel_CG14156
- Chromosome Location
-
Chr.3L: 10243320 - 10244706 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036078
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079294.3 |
NP_524018.2 |
3 - 4 |
1159 |
101 |
View Full Product Details