Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- caup
- Entrez Gene ID
-
39440
- Gene Name
- caupolican
- Gene Aliases
-
CG10605, Caup, Dmel\CG10605, Dmel_CG10605, IRO-C, Iro, Iro-C, IroC, iro, iro-C
- Chromosome Location
-
Chr.3L: 12609631 - 12621798 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015919
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079322.3 |
NP_524046.2 |
1 - 2 |
1189 |
76 |
GenBank mRNA |
BT021246.1 |
- |
1 - 2 |
1186 |
76 |
View Full Product Details