Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
st
Entrez Gene ID
39836
Gene Name
scarlet
Gene Aliases
CG4314, Dmel\CG4314, Dmel_CG4314
Chromosome Location
Chr.3L: 16497651 - 16500460 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003515
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079384.5 NP_524108.1 3 - 4 606 60
GenBank mRNA BT150167.1 - 3 - 4 603 60

View Full Product Details