Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grd
Entrez Gene ID
39984
Gene Name
Glycine receptor
Gene Aliases
CG7446, CT22917, DmGABA(GRD), Dmel\CG7446, Dmel_CG7446, GRD
Chromosome Location
Chr.3L: 17823333 - 17839767 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001134
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079407.3 NP_524131.1 2 - 3 1312 82
GenBank mRNA BT126369.1 - 2 - 3 1312 82
X78349.1 - - 750 82

View Full Product Details