Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
not
Entrez Gene ID
40030
Gene Name
non-stop
Gene Aliases
CG4166, Dmel\CG4166, Dmel_CG4166, NOT, Nonstop, Not, Q9VVR1, UBP8, USP22, Upb8, anon-WO0257455.9, dNonStop, i155, l(3)02069, l(3)j9A6, nonstop
Chromosome Location
Chr.3L: 18618305 - 18622129 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013717
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300177.1 NP_001287106.1 4 - 5 2086 134
NM_079416.4 NP_524140.2 4 - 5 2086 134

View Full Product Details