Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ac78C
Entrez Gene ID
40333
Gene Name
Adenylyl cyclase 78C
Gene Aliases
AC 78C, AC78C, CG10554, CG10564, DAC78C, DAC78C-s, Dmel\CG10564, Dmel_CG10564
Chromosome Location
Chr.3L: 21136447 - 21171930 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024150
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259928.2 NP_001246857.1 3 - 4 423 65
NM_001275208.1 NP_001262137.1 9 - 10 2222 65
NM_001275209.1 NP_001262138.1 10 - 11 2433 65
NM_001275210.1 NP_001262139.1 10 - 11 2430 65
NM_079470.5 NP_524194.3 10 - 11 2433 65

View Full Product Details