Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nrm
Entrez Gene ID
40515
Gene Name
neuromusculin
Gene Aliases
CG32458, CG41452, CG43079, CG8779, CT16082, Dmel\CG43079, Dmel_CG43079
Chromosome Location
Chr.3L: 22998372 - 23040300 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043156.3 NP_001036621.1 11 - 12 2069 167
NM_001259961.2 NP_001246890.1 11 - 12 2069 167
NM_001259962.2 NP_001246891.1 11 - 12 2069 167
NM_001259964.2 NP_001246893.1 11 - 12 2069 167
NM_001275303.1 NP_001262232.1 11 - 12 2069 167
NM_079494.5 NP_524218.4 11 - 12 2069 167
GenBank mRNA BT150396.1 - 11 - 12 1887 167

View Full Product Details