Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cpr50Cb
- Entrez Gene ID
-
36525
- Gene Name
- Cuticular protein 50Cb
- Gene Aliases
-
CG6305, DmelCpr50Cb, Dmel\CG6305, Dmel_CG6305
- Chromosome Location
-
Chr.2R: 13794777 - 13805231 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033869
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137057.3 |
NP_610901.1 |
3 - 4 |
384 |
60 |
GenBank mRNA |
AY060758.1 |
- |
3 - 4 |
358 |
60 |
View Full Product Details