Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
conv
Entrez Gene ID
36588
Gene Name
convoluted
Gene Aliases
ALS, Als, CG8561, CT24963, Dmel\CG8561, Dmel_CG8561, dALS, l(2)k06507b, tartan/capricious-like
Chromosome Location
Chr.2R: 14270729 - 14275495 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261269
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299487.1 NP_001286416.1 6 - 7 2197 79
NM_137104.3 NP_610948.2 6 - 7 2438 79
GenBank mRNA EU814872.1 - 6 - 7 1997 79

View Full Product Details