Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpr51A
Entrez Gene ID
36613
Gene Name
Cuticular protein 51A
Gene Aliases
CG10112, Dmel\CG10112, Dmel_CG10112
Chromosome Location
Chr.2R: 14512559 - 14515211 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033942
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137124.3 NP_610968.1 1 - 2 148 72
GenBank mRNA AY070997.1 - 1 - 2 145 72
KX531580.1 - 1 - 2 65 72

View Full Product Details