Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spoon
Entrez Gene ID
31459
Gene Name
spoonbill
Gene Aliases
AKAP Yu, BcDNA:GM04319, CG3249, Dmel\CG3249, Dmel_CG3249, PKAAP, Spoon, Yu, mdi, yu
Chromosome Location
Chr.X: 5412386 - 5420867 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263987
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167034.2 NP_726992.2 1 - 2 143 66
GenBank mRNA BT099605.1 - 1 - 2 139 66

View Full Product Details