Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cdc7
- Entrez Gene ID
-
31598
- Gene Name
- Cdc7 kinase
- Gene Aliases
-
CG32742, CG8655, CG8656, CT25076, Dmel\CG32742, Dmel_CG32742, L(1)G0148, anon-WO03040301.250, cdc7, cdc7-related, l(1)G0148, l(1)G0149, l(1)G0461
- Chromosome Location
-
Chr.X: 6654541 - 6659709 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028360
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167080.2 |
NP_727103.1 |
2 - 3 |
773 |
69 |
View Full Product Details