Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crag
Entrez Gene ID
31800
Gene Name
Calmodulin-binding protein related to a Rab3 GDP/GTP exchange protein
Gene Aliases
CG12737, CJ, CRAG, Dmel\CG12737, Dmel_CG12737
Chromosome Location
Chr.X: 8592031 - 8602036 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025864
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169239.2 NP_001162710.2 12 - 13 5090 58
NM_080341.4 NP_525080.3 12 - 13 5090 58
NM_167164.4 NP_727281.2 11 - 12 5009 58
GenBank mRNA BT031006.1 - 10 - 11 4457 58

View Full Product Details