Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PlexB
Entrez Gene ID
43766
Gene Name
Plexin B
Gene Aliases
CG17245, Dmel\CG17245, Dmel_CG17245, PLEXB, Plex B, plex, plex B, plexB
Chromosome Location
Chr.4: 32478 - 43778 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025740
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258471.2 NP_001245400.1 3 - 4 6217 105
NM_079877.4 NP_524616.2 3 - 4 6217 105
GenBank mRNA BT010009.1 - 3 - 4 6216 105

View Full Product Details