Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Lip2
- Entrez Gene ID
-
43980
- Gene Name
- Lipase 2
- Gene Aliases
-
CG17116, DILP2, Dmel\CG17116, Dmel_CG17116, Dmlip2, dilp2, lip2
- Chromosome Location
-
Chr.2L: 10701940 - 10709453 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024740
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079928.2 |
NP_524667.1 |
1 - 2 |
114 |
66 |
View Full Product Details