Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP1
Entrez Gene ID
43981
Gene Name
DISCO Interacting Protein 1
Gene Aliases
CG17686, Dmel\CG17686, Dmel_CG17686, TO34, TO67, UbxBP1, clot 10495, clot 2020, dip1, dip1a, klt
Chromosome Location
Chr.X: 21624796 - 21630148 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024807
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169343.2 NP_001162814.1 2 - 3 2368 56
NM_001169344.2 NP_001162815.1 4 - 5 2094 56
NM_001169345.2 NP_001162816.1 3 - 4 1011 56
NM_001169346.2 NP_001162817.1 4 - 5 2022 56
NM_079929.3 NP_524668.2 3 - 4 1104 56
NM_167772.2 NP_728449.1 3 - 4 1149 56
NM_167773.3 NP_728450.1 4 - 5 2098 56
NM_167774.3 NP_728451.2 4 - 5 2053 56
GenBank mRNA AF175711.2 - 3 - 4 1132 56
AF175713.1 - - 995 56
AY121664.1 - 4 - 5 2104 56
BT021433.1 - 4 - 5 2097 56
BT099548.1 - 3 - 4 1127 56
BT125869.1 - 3 - 4 1068 56

View Full Product Details