Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bgm
Entrez Gene ID
44117
Gene Name
bubblegum
Gene Aliases
2a9hom, BG:DS01514.2, BG:DS0514.2, Bgm, CG4501, Dmel\CG4501, Dmel_CG4501
Chromosome Location
Chr.2L: 14013004 - 14019328 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027348
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298994.1 NP_001285923.1 5 - 6 1824 66
NM_079959.4 NP_524698.1 5 - 6 1861 66

View Full Product Details