Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ETH
- Entrez Gene ID
-
44125
- Gene Name
- Ecdysis triggering hormone
- Gene Aliases
-
CG18105, DmETH, Dmel\CG18105, Dmel_CG18105, Drm-ETH-1, Drm-ETH-2, ETH-1, ETH-2, ETH1, ETH2, Eth, eth
- Chromosome Location
-
Chr.2R: 24794670 - 24795482 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028738
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079960.4 |
NP_524699.1 |
1 - 2 |
207 |
65 |
GenBank mRNA |
BT120013.1 |
- |
1 - 2 |
183 |
65 |
View Full Product Details