Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hbs
Entrez Gene ID
44129
Gene Name
hibris
Gene Aliases
CG7449, CT22841, Dmel\CG7449, Dmel_CG7449, Hbs, icon
Chromosome Location
Chr.2R: 15011396 - 15042134 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029082
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299510.1 NP_001286439.1 3 - 4 1134 105
NM_079963.4 NP_524702.3 3 - 4 1134 105
NM_176175.2 NP_788355.1 3 - 4 1134 105
GenBank mRNA AY121698.1 - 3 - 4 1132 105

View Full Product Details