Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Picot
Entrez Gene ID
36865
Gene Name
CG8098-PA
Gene Aliases
CG8098, Dmel\CG8098, Dmel_CG8098, MFS8, picot
Chromosome Location
Chr.2R: 16655323 - 16667675 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024315
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299567.1 NP_001286496.1 1 - 2 449 77
NM_001299568.1 NP_001286497.1 3 - 4 952 77
NM_137302.3 NP_611146.1 2 - 3 817 77
GenBank mRNA AY069501.1 - 2 - 3 807 77

View Full Product Details