Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cht8
- Entrez Gene ID
-
37390
- Gene Name
- Chitinase 8
- Gene Aliases
-
CG9357, DmCht8, Dmel\CG9357, Dmel_CG9357
- Chromosome Location
-
Chr.2R: 21061646 - 21063212 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034580
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137698.1 |
NP_611542.2 |
2 - 3 |
542 |
96 |
View Full Product Details