Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smr
Entrez Gene ID
32225
Gene Name
Smrter
Gene Aliases
CG12719, CG4013, Dmel\CG4013, Dmel_CG4013, E52, SMR, SMRTER, anon-WO0153538.73, l(1)G0060, l(1)G0361, lincRNA.983, smr
Chromosome Location
Chr.X: 12684203 - 12742618 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265523
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272562.1 NP_001259491.1 6 - 7 5763 77
NM_080536.4 NP_536797.3 5 - 6 5144 77
NM_167334.2 NP_727634.2 5 - 6 5058 77
NM_167335.2 NP_727635.2 6 - 7 5823 77

View Full Product Details