Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smr
Entrez Gene ID
32225
Gene Name
Smrter
Gene Aliases
CG12719, CG4013, Dmel\CG4013, Dmel_CG4013, E52, SMR, SMRTER, anon-WO0153538.73, l(1)G0060, l(1)G0361, lincRNA.983, smr
Chromosome Location
Chr.X: 12684203 - 12742618 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265523
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272562.1 NP_001259491.1 7 - 8 6171 73
NM_080536.4 NP_536797.3 6 - 7 5552 73
NM_167334.2 NP_727634.2 6 - 7 5466 73
NM_167335.2 NP_727635.2 7 - 8 6231 73

View Full Product Details