Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mamo
Entrez Gene ID
32353
Gene Name
maternal gene required for meiosis
Gene Aliases
CG11071, CG11072, CG11075, CG11082, CG32606, CG32611, CG34346, CG44132, CG45477, CR44656, Dmel\CG45477, Dmel_CG45477, lincRNA.990
Chromosome Location
Chr.X: 13846116 - 13990527 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267033
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201660.3 NP_001188589.1 7 - 8 3990 82
NM_001258732.2 NP_001245661.1 8 - 9 4917 82
NM_001258733.1 NP_001245662.1 7 - 8 4041 82
NM_001272610.2 NP_001259539.1 9 - 10 4865 82

View Full Product Details