Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nipsnap
Entrez Gene ID
32573
Gene Name
CG9212-PA
Gene Aliases
CG9212, Dmel\CG9212, Dmel_CG9212
Chromosome Location
Chr.X: 16070365 - 16073568 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030724
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298387.1 NP_001285316.1 1 - 2 252 89
NM_132875.4 NP_573103.1 2 - 3 236 89
NM_167483.3 NP_727931.1 2 - 3 277 89
NM_167484.3 NP_727932.1 3 - 4 158 89
GenBank mRNA AY061048.1 - 2 - 3 169 89

View Full Product Details