Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp19a
- Entrez Gene ID
-
33037
- Gene Name
- Odorant-binding protein 19a
- Gene Aliases
-
19a, CG11748, DmelObp19a, Dmel\CG11748, Dmel_CG11748, OBP19a
- Chromosome Location
-
Chr.X: 20415744 - 20416521 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031109
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167700.3 |
NP_728338.2 |
1 - 2 |
96 |
71 |
View Full Product Details