Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk12
Entrez Gene ID
37566
Gene Name
pickpocket 12
Gene Aliases
CG10972, Dmel\CG10972, Dmel_CG10972, PPK12
Chromosome Location
Chr.2R: 22427479 - 22429584 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034730
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137828.1 NP_611672.1 6 - 7 1424 69
GenBank mRNA AY226543.1 - 6 - 7 1424 69

View Full Product Details