Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITP
Entrez Gene ID
37921
Gene Name
Ion transport peptide
Gene Aliases
BcDNA:SD05282, CG13586, CHH-like, Dmel\CG13586, Dmel_CG13586, Drm-ITP, DrmITP, DrmITPL1, DrmITPL2, Itp, chgh, itp
Chromosome Location
Chr.2R: 24537421 - 24558961 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035023
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169823.2 NP_001163294.1 3 - 4 878 73
NM_001299916.1 NP_001286845.1 2 - 3 321 73
NM_138087.6 NP_611931.3 2 - 3 1299 73
GenBank mRNA AY060468.1 - 2 - 3 1275 73
KX531271.1 - 1 - 2 219 73

View Full Product Details