Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
deltaCOP
Entrez Gene ID
45250
Gene Name
Coat Protein (coatomer) delta
Gene Aliases
63B12S, ARCN1, Arch, CG14813, DM63B12S, Delta COP, DeltaCop, Dmel\CG14813, Dmel_CG14813, EG:63B12.10, ESTS:63B12S, anon-WO03040301.246, delta COP, delta-Cop, deltaCop, l(1)G0051, l(1)G0301, l(1)G0450
Chromosome Location
Chr.X: 1859141 - 1861790 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028969
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169171.2 NP_001162642.1 6 - 7 1384 99
NM_143755.4 NP_652012.1 6 - 7 1387 99
GenBank mRNA AY051528.1 - 6 - 7 1389 99

View Full Product Details