Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mei-P26
Entrez Gene ID
45775
Gene Name
meiotic P26
Gene Aliases
AF145661, BcDNA:GH10646, CG12218, Dmel\CG12218, Dmel_CG12218, MEI-P26, Mei-P26, mei-p26, meiP26
Chromosome Location
Chr.X: 9163255 - 9189309 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026206
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272447.2 NP_001259376.1 4 - 5 3585 80
NM_001272448.2 NP_001259377.1 5 - 6 4046 80
NM_001272449.1 NP_001259378.1 5 - 6 4023 80
NM_001298110.1 NP_001285039.1 4 - 5 3621 80
NM_001369966.1 NP_001356936.1 5 - 6 3887 80
NM_143765.4 NP_652022.1 5 - 6 3887 80
GenBank mRNA BT025212.1 - 5 - 6 3890 80

View Full Product Details