Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pgant35A
- Entrez Gene ID
-
48775
- Gene Name
- Polypeptide N-acetylgalactosaminyltransferase 35A
- Gene Aliases
-
A3, BG:DS01068.8, CG7480, Dmel\CG7480, Dmel_CG7480, GalNAc-T, br12, dGalNAc-T1, l(2)35Aa, l(2)br12, l35Aa, pgant35, pgant35A
- Chromosome Location
-
Chr.2L: 14359252 - 14361411 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001970
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143812.4 |
NP_652069.2 |
1 - 1 |
2016 |
100 |
GenBank mRNA |
BT128701.1 |
- |
1 - 1 |
1944 |
100 |
View Full Product Details