Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZnT63C
Entrez Gene ID
38407
Gene Name
Zinc transporter 63C
Gene Aliases
CG17723, Dmel\CG17723, Dmel_CG17723, ZNT63C, ZnT1, cg17723, dZNT1, dZNT63C, dZnT1, dZnT63C
Chromosome Location
Chr.3L: 3303708 - 3314009 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035432
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274435.1 NP_001261364.1 7 - 8 1373 66
NM_001274436.1 NP_001261365.1 6 - 7 1299 66
NM_139544.3 NP_647801.1 6 - 7 1309 66
NM_168015.2 NP_728856.1 7 - 8 1373 66
NM_168016.2 NP_728857.1 7 - 8 1542 66
NM_168017.2 NP_728858.1 7 - 8 1794 66
NM_168018.2 NP_728859.1 6 - 7 1295 66
GenBank mRNA AY058521.1 - 6 - 7 1267 66

View Full Product Details