Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snz
Entrez Gene ID
31704
Gene Name
snazarus
Gene Aliases
CG1514, DmSNX25, Dmel\CG1514, Dmel_CG1514, SNZ, Snz
Chromosome Location
Chr.X: 7912089 - 7918444 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029976
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298064.1 NP_001284993.1 4 - 5 1441 65
NM_132193.2 NP_572421.1 4 - 5 1441 65
GenBank mRNA BT150246.1 - 4 - 5 1208 65

View Full Product Details