Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tudor-SN
Entrez Gene ID
38045
Gene Name
Tudor staphylococcal nuclease
Gene Aliases
CG7008, Dmel\CG7008, Dmel_CG7008, SND1, TSN, TSN-1, TUDOR-SN, dP100, p100, tudor-SN
Chromosome Location
Chr.3L: 259043 - 264380 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035121
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274266.1 NP_001261195.1 2 - 3 1105 54
NM_138177.3 NP_612021.1 2 - 3 1105 54
GenBank mRNA BT003270.1 - 2 - 3 1079 54

View Full Product Details