Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RabX5
- Entrez Gene ID
-
38209
- Gene Name
- CG7980-PB
- Gene Aliases
-
CG7980, DmRabX5, Dmel\CG7980, Dmel_CG7980, Q9W0A7, rabX5
- Chromosome Location
-
Chr.3L: 1668901 - 1670108 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035255
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139387.3 |
NP_647644.2 |
1 - 2 |
274 |
68 |
View Full Product Details