Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
armi
Entrez Gene ID
38427
Gene Name
armitage
Gene Aliases
APC16, ARMI, Armi, CG11513, Dmel\CG11513, Dmel_CG11513
Chromosome Location
Chr.3L: 3461317 - 3466314 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041164
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014556.4 NP_001014556.1 2 - 3 279 69

View Full Product Details