Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ccz1
- Entrez Gene ID
-
38455
- Gene Name
- Caffeine, calcium, zinc sensitivity 1
- Gene Aliases
-
C7orf28B, CG14980, Dccz1, Dmel\CG14980, Dmel_CG14980, NP_647835
- Chromosome Location
-
Chr.3L: 3897444 - 3899156 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035470
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139578.3 |
NP_647835.1 |
2 - 3 |
234 |
65 |
GenBank mRNA |
AY071668.1 |
- |
2 - 3 |
235 |
65 |
View Full Product Details