Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ida
- Entrez Gene ID
-
45574
- Gene Name
- imaginal discs arrested
- Gene Aliases
-
APC5, Apc5, CG10850, Dmel\CG10850, Dmel_CG10850, IDA, Ida, l(3)63Fb, l(3)SH7
- Chromosome Location
-
Chr.3L: 3905344 - 3907813 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041147
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139580.3 |
NP_647837.2 |
1 - 2 |
184 |
85 |
View Full Product Details