Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adi1
Entrez Gene ID
326189
Gene Name
Acireductone dioxygenase 1
Gene Aliases
CG32068, CG8000, Dadi1, Dmel\CG32068, Dmel_CG32068, adi1
Chromosome Location
Chr.3L: 10663376 - 10664435 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052068
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104107.2 NP_001097577.1 2 - 3 274 64

View Full Product Details