Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nc73EF
Entrez Gene ID
39899
Gene Name
Neural conserved at 73EF
Gene Aliases
141810_at, CG11661, DOGDH, Dmel\CG11661, Dmel_CG11661, E1, Nc73eF, anon-WO0118547.329, dOgdh
Chromosome Location
Chr.3L: 16954822 - 16968724 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010352
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_168701.5 NP_730223.1 10 - 11 3037 91
NM_168702.2 NP_730224.1 10 - 11 2998 91
NM_168703.5 NP_730225.1 10 - 11 3040 91
NM_176340.3 NP_788518.1 10 - 11 3018 91
NM_176341.3 NP_788519.1 10 - 11 2991 91
NM_176342.5 NP_788520.2 10 - 11 3042 91
GenBank mRNA BT003743.1 - 10 - 11 3018 91
BT012466.1 - 10 - 11 3005 91

View Full Product Details