Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ome
- Entrez Gene ID
-
44297
- Gene Name
- omega
- Gene Aliases
-
BEST:LD21715, CG17705, CG32143, CG32145, CG42280, CG9370, Dmel\CG42280, Dmel_CG42280, LD21715, Ome
- Chromosome Location
-
Chr.3L: 14672839 - 14747868 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259175
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080000.4 |
NP_524739.1 |
2 - 3 |
216 |
72 |
GenBank mRNA |
AY058514.1 |
- |
2 - 3 |
189 |
72 |
View Full Product Details