Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rph
Entrez Gene ID
32002
Gene Name
Rabphilin
Gene Aliases
CG11556, Dmel\CG11556, Dmel_CG11556, rph
Chromosome Location
Chr.X: 10749077 - 10757666 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030230
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169244.1 NP_001162715.1 6 - 7 2319 68
NM_001272499.2 NP_001259428.1 6 - 7 2452 68
NM_001272500.1 NP_001259429.1 5 - 6 2151 68
NM_132423.2 NP_572651.1 4 - 5 1804 68
GenBank mRNA BT015226.1 - 6 - 7 2319 68

View Full Product Details