Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ir10a
Entrez Gene ID
32067
Gene Name
Ionotropic receptor 10a
Gene Aliases
CG12623, CG12624, CG34143, DmelIR10a, Dmel\CG34143, Dmel_CG34143, IR10a
Chromosome Location
Chr.X: 11308533 - 11311208 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0083979
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103479.1 NP_001096949.1 1 - 1 567 77

View Full Product Details