Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir11a
- Entrez Gene ID
-
32189
- Gene Name
- Ionotropic receptor 11a
- Gene Aliases
-
CG15732, CT35969, DmelIR11a, Dmel\CG15732, Dmel_CG15732, IR11a, Q9VYN4_DROME
- Chromosome Location
-
Chr.X: 12375596 - 12377755 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030385
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132567.1 |
NP_572795.2 |
2 - 3 |
1000 |
81 |
View Full Product Details