Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fne
Entrez Gene ID
32245
Gene Name
found in neurons
Gene Aliases
BEST:HL02010, BcDNA:HL02010, BcDNA:RE25290, CG4396, CR32646, Dmel\CG4396, Dmel_CG4396, FNE, Fne, MRE28, cg4396, lincRNA.985
Chromosome Location
Chr.X: 12917477 - 12950417 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086675
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103495.5 NP_001096965.1 4 - 5 1571 68
NM_001258717.1 NP_001245646.1 5 - 6 3091 68
NM_001258718.3 NP_001245647.1 5 - 6 2876 68
NM_001258719.2 NP_001245648.1 4 - 5 2657 68
NM_001258720.3 NP_001245649.2 3 - 4 1903 68
NM_001272568.2 NP_001259497.1 3 - 4 1363 68
NM_001272569.1 NP_001259498.1 4 - 5 1829 68
NM_001298244.1 NP_001285173.1 3 - 4 1354 68
NM_132614.6 NP_572842.1 3 - 4 1912 68
GenBank mRNA BT120384.2 - 2 - 3 248 68
BT125663.1 - 2 - 3 239 68

View Full Product Details