Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rad
Entrez Gene ID
32253
Gene Name
radish
Gene Aliases
CG15720, CG18646, CG42628, CG42629, CG4346, CG4368, CG44424, Dmel\CG44424, Dmel_CG44424, FBpp0073550, R5_X:12971309..12973132, Rad, Rsh, rsh
Chromosome Location
Chr.X: 12988615 - 13079070 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265597
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298247.1 NP_001285176.1 14 - 15 4090 76
NM_001298248.1 NP_001285177.1 14 - 15 4314 76
NM_132622.4 NP_572850.5 10 - 11 2502 76

View Full Product Details