Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dob
Entrez Gene ID
32436
Gene Name
doppelganger von brummer
Gene Aliases
CG5560, Dmel\CG5560, Dmel_CG5560, Dob
Chromosome Location
Chr.X: 15067007 - 15069091 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030607
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132767.3 NP_572995.1 1 - 2 652 65
GenBank mRNA AY069470.1 - 1 - 2 595 65

View Full Product Details