Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tay
Entrez Gene ID
32547
Gene Name
tay bridge
Gene Aliases
CG9056, Dmel\CG9056, Dmel_CG9056, XI, dAUTS2, l(1)13Ff, l(1)14Ac, l(1)XI
Chromosome Location
Chr.X: 15876368 - 15887311 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260938
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272656.2 NP_001259585.1 11 - 12 6019 65
NM_001272657.2 NP_001259586.1 11 - 12 5939 65
NM_001298376.1 NP_001285305.1 11 - 12 6041 65
NM_132858.3 NP_573086.2 11 - 12 6044 65
GenBank mRNA AY118524.1 - 11 - 12 6019 65

View Full Product Details