Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP-delta
Entrez Gene ID
5740816
Gene Name
Dpr-interacting protein delta
Gene Aliases
34391, CG15214, CG34391, CG4814, CT15465, CT35147, Dmel\CG34391, Dmel_CG34391
Chromosome Location
Chr.3L: 5424109 - 5442399 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085420
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104038.4 NP_001097508.2 7 - 8 1328 92
NM_001274512.2 NP_001261441.1 7 - 8 1328 92

View Full Product Details