Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Lkr
- Entrez Gene ID
-
38622
- Gene Name
- Leucokinin receptor
- Gene Aliases
-
CG10626, DLKR, Dmel\CG10626, Dmel_CG10626, LK-R, LKR, LkR, anon-WO0131005.17, anon-WO0170980.118, anon-WO0170980.119, dLKR, lkr
- Chromosome Location
-
Chr.3L: 5512421 - 5529341 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035610
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139711.3 |
NP_647968.3 |
6 - 7 |
1309 |
82 |
View Full Product Details